VWR® Glass Joint Clamps, PTFE
Fournisseur: VWR International
CE Compliant
Certificats
A propos de cet article
PTFE joint clamps can withstand temperatures of up to 260°C (500°F).
- Chemically resistant and inert
Feature PTFE-encapsulated steel springs that can be adjusted by hand.
Customers Who Bought This Also Bought