Standard Taper Clamps, Wheaton
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificats
A propos de cet article
Color-coded clamps hold conical ground joints secure under pressure.
Molded from acetal plastic, clamps are safe for use to 150°C (302°F). Unlike steel wire clamps, these will not scratch glass or bend out of shape. Available in the four most commonly used sizes; assorted pack 21734-920 includes six of each size/color.
Customers Who Bought This Also Bought