ContactUsLinkComponent
Water Test Comparator and Color Disc Test Kits, Lovibond®, Tintometer
Water Test Comparator and Color Disc Test Kits, Lovibond®, Tintometer
# de catalogue 66146-153
Fournisseur:  Lovibond
Small Business Enterprise
Numéro CAS:  
Water Test Comparator and Color Disc Test Kits, Lovibond®, Tintometer
# de catalogue 66146-153
Fournisseur:  Lovibond
Small Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Chemical method:
    DPD (N,N-diethyl-p-phenylenediamine)
  • Description:
    DPD No. 3 Tablets
  • Cat. no.:
    66146-153
  • Supplier No.:
    433-IT
Spécifications
A propos de cet article
Ideal for testing water, wastes and process fluids in the lab, plant, classroom or field
Color Disc Test Kits for use with Comparator contain color disc, stable reagents for tests, dispensers, labware and easy step-by-step instructions. Color discs are non-fading and will continue to give accurate results even after prolonged exposure to sunlight, temperature extremes, excessive humidity and corrosive fumes. Chorine kits supply sufficient reagents for 50 tests; pH kits, enough for 120 tests.
Chlorine tests for free, combined and total chlorine are acceptable by EPA for NIPDWR compliance monitoring.
The comparator accepts interchangeable discs containing 9 or 10 permanent color standards. A touch of the finger brings one standard after another into view for comparison with the color of the reagent-treated sample. When the colors match, the correct test result appears in an aperture in the comparator. The magnifying prism enhances the comparison process by bringing colors side-by-side into one enlarged image for a positive color match. A precision diffuser provides equal light to the color standard and sample. The 13mm square tube shows uniform color from edge to edge. Pocket size: 9.2Wx9.2Hx3.8Dcm (35/8x35/8x11/2"). Chemical-resistant phenolic construction. Optional “Daylite” Illuminator provides correct color matching conditions at all times and locations, irrespective of lighting conditions. Features a color-correcting lamp and filter in a sloping-front housing.
Ordering information: Supplied with two precision square tubes. Comparator may be ordered with or without magnifying prism. Order Illuminator and Color Disc Test Kits separately.
Customers Who Bought This Also Bought