Pura-Fit® 6800 and 6900 Earplugs, Moldex®
Fournisseur: Moldex/Metric
Certificats
A propos de cet article
For convenient, comfortable hearing protection
- Convenient, comfortable hearing protection
- Soft foam fits snugly, tapered design allows easy insertion
- Brightly colored for easy compliance checks
Earplugs 56610-663 are hygienically pair-packed in biodegradable paper pouches. Earplugs with cord (56610-673) are packaged in a plastic pouch.
Soft foam plugs fit snugly and comfortably into ears. Tapered design allows easy insertion into any size ear canal. Brightly colored for easy compliance checks.
Certifications: Meet ANSI S3.19 in independent testing.
Spécifications
- Catalog No:
- 56610-663
- 56610-673
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health