Stainless Steel Powder Feeders, Tecniplast®
Fournisseur: Tecniplast Usa
Certificats
A propos de cet article
Designed to be hung from the inner lip of size D cages, providing an ideal outlet for powdered feed studies while reducing waste
The two-part feeder consists of a lower tray/hopper that is filled with powdered feed and an upper lid/divider that slides directly onto the food. A series of stainless steel crosswires on the top section provides multiple slots through which the animals feed, encouraging the consumption of fine particles.
Feeders are created with multiple partitions to allow simultaneous access by two or more animals. The small compartment size prevents animals from entering and nesting.
Customers Who Bought This Also Bought