Vee Gee Graduated Cylinders, Student Grade
Supplier: Heathrow Scientific
Certificates
About this item
These high-quality graduated cylinders are made of premium borosilicate glass and comply with ASTM E-1272 standards, guaranteeing your investment in high-quality glassware.
- Made out of high-quality borosilicate glass
- Double metric scale
- Graduated 'to contain'
- Contains durable white enamel graduation markings
- Feature hexagonal bases for added stability
- All sizes come with a protective bumper guard
- Calibration temperature: 20 °C (68 °F)
With their exceptional quality, precise accuracy, and ease of use, they are a great tool to use for all of your liquid handling needs.
Each graduated cylinder features a double metric scale, which is calibrated to contain, ensuring that the results of your experiments are precise and trustworthy. These graduated cylinders also feature an easy-pour spout, which allows for a controlled and drip-free pour, preventing spillage. All sizes have a hexagonal base for added stability and contain white enamel graduation markings that are easy to read, minimising measurement errors.
Certifications: Exceeds ASTM E-1272 standards.
Frequently Bought Together