Vee Gee Glass Burettes with PTFE Stopcock
Fournisseur: Heathrow Scientific
Certificats
A propos de cet article
Elevate your titration processes with these premium glass burettes. These burettes provide a clear view of your titration processes, ensuring reliability in volumetric determinations.
- Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination
- Designed 'to deliver'
- Made from high-quality borosilicate tubing with a low coefficient of expansion
- Feature permanent graduation markings that are large and easy-to-read
Each burette is made from high-quality borosilicate glass tubing with a low coefficient of expansion and comes with a PTFE stopcock, which helps to eliminate freezing and lubricant contamination. They also feature permanent graduation markings that are large and easy to read.
Certifications: Meet ASTM E-287 Class A specifications.
Customers Who Bought This Also Bought