Animal Water Bottles, Tecniplast®
# de catalogue 75876-810
Fournisseur: Tecniplast Usa
Spécifications
- Volume:600 ml
- Color:Clear
- Description:Polycarbonate Bottles
- Cat. no.:75876-810
- Supplier No.:ACBT0702
Spécifications
A propos de cet article
Designed for use in animal housing and feeding applications, water bottles feature wide mouths to aid in filling and emptying, and embossed, molded graduations for accurate measurement
Manufactured with consistent thickness and rigid walls, bottles remain rugged and shatter-proof even when autoclaved repeatedly. Available in multiple materials and clear or amber hues, they are also equipped with silicone neck rings to facilitate the placing and removal of caps. Bottles 95042-952 and 95057-872 feature a cone neck. Supplied without caps. Autoclavable at 121°C (250°F) for 20 minutes.
Customers Who Bought This Also Bought