Rodent Retreats Rat Enrichment Devices, Bio-Serv
Fournisseur: Bio-Serv
Certificats
A propos de cet article
Polycarbonate devices offer rodents a place to rest comfortably on a solid surface, rather than on uncomfortable wire-bottom caging commonly used in many toxicology or other studies
Enhance animal well-being while minimizing stress and cage-associated foot lesions. Animals are easily observed through the transparent sides. Devices do not subtract from cage-space requirements and stack easily for cleaning. Cage washer-safe and autoclavable. May be used for aquatics. Certified (contaminant screened).
Customers Who Bought This Also Bought