Claisen Distillation Adapters, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Adapters feature a thermometer neck on the vertical side tube for immersion thermometers
The inner drip tip joint on the sidearm and the top outer joint and lower inner joint on the vertical tube are all the same [ST] joint size.
Customers Who Bought This Also Bought