Isopropyl Alcohol Cleaner, Stoner
# de catalogue 89065-938
Fournisseur: Stoner
Spécifications
- Description:Pail
- Size:19 L (5 gal.)
- Cat. no.:89065-938
- Supplier No.:D005
Spécifications
A propos de cet article
Pure isopropyl alcohol acts as a defluxer and general-purpose degreaser to remove dirt, oxides, oils, smoke film, and corrosion from metal and plastic surfaces.
- IPA is 99.9% pure
- Evaporates fast
- Leaves no residue
Apply with squirt bottles, pump spray dispensers, swabs, wipes, or tank dipping techniques.
Customers Who Bought This Also Bought