Vee Gee Volumetric Flasks with Glass Stopper
Supplier: Heathrow Scientific
Certificates
About this item
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers.
- Made of premium borosilicate glass with a low coefficient of expansion
- Designed 'to contain' with a temperature of calibration at 20 °C (68 °F)
- The graduation line is sharply defined and of uniform width
- Each flask has a large marking square and includes a glass stopper
- Wide range of sizes available (5 to 2000 ml)
Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions. Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.
Certifications: Meets ASTM E-288 Class A specifications and USP standards for glassware.
Customers Who Bought This Also Bought