DECON-SPORE 200 Plus®, Veltek Associates
Supplier: Veltek Associates
Certificates
About this item
Designed for the sterilization of manufacturing, packaging, and filling equipment in aseptic processing areas.
- Prefiltered to 0.2 um
- Completely traceable and validated for sterility and shelf life
- Effective disinfectant in water up to 400ppm hardness in the presence of 5% serum contamination and dried soap film residue
- Avaliable in three configurations: concentrate, disinfectant use dilution, and sporicidal use dilution."
Certifications: Delivered with lot specific Certificate of Analysis and Certificate of Sterility.
Packaging: Double bagged in easy tear bag.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.