A propos de cet article
The Opti-Swab® liquid amies transport system is ideal for the collection and transport of aerobic, anaerobic, and fastidious bacteria. It is also suitable for use in bacterial culture, gram staining, rapid tests, automation, and molecular tests when validated by test manufacturers and/or laboratory.
- Ready-to-use liquid amies collection and transport system
- For the collection and transport of aerobic, anaerobic, and fastidious bacteria
- Suitable for use in bacterial culture, gram staining, rapid tests, automation and molecular tests when validated by test manufacturers and/or laboratory
- Advanced kit design is easy to use and reliable
- Self-centering cap is leak-proof and facilitates easy swab removal and disposal
- Soft tip for patient comfort
- Premium medical grade plastic components to improve product shelf life
- Shelf life: 18 months
- For In Vitro diagnostic use
- Made in USA
High-performance swab tips are available in three formats. The HydraFlock swab (3317-H) with 100 mm molded breakpoint is designed to accommodate nasopharyngeal (NP) sampling sites. The HydraFlock swab (3316-H) with molded breakpoint is designed to accommodate eye, ear, urogenital, and pediatric sampling sites. The HydraFlock swab (3506-H) with molded breakpoint is designed to accommodate nose, throat, vaginal, rectal, and wound sampling sites.