Rhino Series Refractometers, Percent Solids (Brix), Reichert
Fournisseur: Reichert Technologies
Certificats
A propos de cet article
These automatic temperature-compensated hand refractometers are read directly and instantly without a water bath
Refractometers are constructed of durable, shock-resistant Xenoy® polycarbonate materials for demanding environments and applications. Water resistant. Dustproof.
No need to make separate temperature readings or refer to conversion tables. The Brix scale is based on percent by weight of sucrose. Useful in monitoring sugar level of soft drinks and juices and to check dissolved solids in ripening fruit crops.
Certifications: IP67 rated for entire unit, including eyepiece.
Spécifications
- Catalog No:
- 55680-369
- 55680-347
- Accuracy:
- ±1 scale division
- ±1 scale division
- Dimensions:
- 23.5L x 3.8W x 3.8H cm (91/4 x 11/2 x 11/2")
- 23.5L x 3.8W x 3.8H cm (91/4 x 11/2 x 11/2")
- Weight:
- 162.4 g (5.7 oz.)
- 162.4 g (5.7 oz.)