VWR® −40 °C Ultra-Low Temperature Freezers
Supplier: VWR International
Certificates
About this item
Dependable –40 °C storage, powered by H-drive for operational savings and sustainability.
- Low energy
- Low peak variation
- Fast door opening recovery
- Long warm up time
- Four sizes
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs. User friendly and quiet.
Certifications: SNAP Compliant.
Ordering information: Options and accessories are sold separately; contact your VWR representative for more information.
Specifications
- Catalog No:
- 76307-990
- 76307-986
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient