Rare Earth Magnet
Catalog # 470356-300
Supplier: AEROLITE METEORITES INC SE
New Product
Specifications
- Description:Rare earth magnet, 1¹/₂׳/₄×¹/₄"
- Cat. no.:470356-300
- Weight:10 g
Specifications
About this item
This magnet is just what you need for testing suspected space rocks in the lab or field.
- 30 lb. capacity
Caution: Not made for children.
Customers Who Bought This Also Bought