Anti-Static Glass Cleaner, Stoner®
# de catalogue 89065-928
Fournisseur: Stoner
Spécifications
- Description:Aerosol Can
- Size:368 g (13 oz.)
- Cat. no.:89065-928
- Supplier No.:A166
Spécifications
A propos de cet article
Static-dissipating cleaner prevents build-up or static charges when used regularly
Cleaner has a fast-acting, non-abrasive, water-based, foaming formula to remove tough dirt, dust, grease, lint, oil, toner, carbon, fingerprints, and smudges from glass, computer or television screens, CRTs, copiers, scanners, windows, and windshields. It can also be used on plastic, vinyl, and painted surfaces. A special foaming action keeps cleaner in place until wiped, so it will not drip or run. Cleaner dries quickly and leaves no streaks, haze, or film.
Customers Who Bought This Also Bought