Invisible Glass Cleaner, Stoner
# de catalogue 89065-936
Fournisseur: Stoner
Spécifications
- Description:Aerosol can
- Size:538 g (19 oz.)
- Cat. no.:89065-936
- Supplier No.:91164
Spécifications
A propos de cet article
Cleaner removes dust, dirt, oil, grease, and fingerprints from windshields, windows, and mirrors without leaving streaks or residue.
- Aerosol formula / contains no soaps, foams, scents or dyes
- Superior cleaning power and invisible clarity
- Cleans windows, mirrors, windscreens and other glass surfaces
- No streak, no haze, no residue formula
Non-abrasive formula evaporates completely.
Customers Who Bought This Also Bought