Overpack and Salvage Drums, UN and DOT, Eagle Mfg
Fournisseur: Eagle Manufacturing
Certificats
A propos de cet article
Designed for use with hazardous materials, Packing Groups I, II, and III.
Constructed of blow-molded high-density polyethylene with UV inhibitors for superior strength. Chemically-resistant to most materials. Safety yellow with removable head. Forkliftable. Nestable for easy storage. Suitable for secondary containment storage, transportation, and cleanup.
- Constructed of blow-molded high-density polyethylene with UV inhibitors for superior strength
- Chemically-resistant to most materials
- Safety yellow with removable head
- Forkliftable
- Nestable for easy storage
- Suitable for secondary containment storage, transportation, and cleanup
Salvage drums have a metal bolt ring locking band and meet 3psi pressure test for Salvage Drum Compliance. Overpack drums have a metal locking band. All drums are UN certified to HM-181. Salvage drums meet DOT 49 CFR 173.3(c). Overpack drums meet DOT 49 CFR 173.25.
Customers Who Bought This Also Bought