KIMAX® Viscosity Tube, Varnish, (Bubble Tube), Kimble Chase, DWK Life Sciences
# de catalogue 52642-000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Varnish Viscosity Tube
- Cat. no.:52642-000
- Supplier No.:46463-1075
Spécifications
A propos de cet article
For rough control work where bubble comparison is made to limit varnish samples of known viscosity, reference ASTM D 1545
114 mm long with 10.75 ±0.125 mm inside diameter. With durable white ceramic enamel lines at 27, 100 and 108 mm from the outside bottom.
Customers Who Bought This Also Bought