Synthware Inlet Adapters with Hose Connection, Kemtech America
# de catalogue 60000-158
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint:24/40
- Cat. no.:60000-158
- Supplier No.:A532440
Spécifications
A propos de cet article
This inlet adapter is designed for N2/Ar introduction or vacuum.
- [ST] joint
- With hose connection
- One nylon cap and one o-ring included
Hose connection is 8 mm. Supplied with one nylon cap and one Viton® O-ring.
Customers Who Bought This Also Bought