EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
For the collection and storage of water samples.
- Made from KG-33 clear borosilicate glass
Clear glass vials have 0.005" PTFE-faced 0.120" silicone rubber septa with open-top screw-caps attached.
Comply with guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603, and 604).
Ordering information: Vials are packaged after the production process in shrink-wrapped modules to help reduce particulate contamination.
Customers Who Bought This Also Bought