VWR® Microwave Reaction Vials
Supplier: VWR International
Certificates
About this item
Vials are manufactured from Type I, Class A (USP Type I) heavy-wall borosilicate glass and are designed to withstand pressures up to 300 psi (20 bar).
- Borosilicate glass fabrication
- Heavy-walled design
Vials are designed to withstand great pressures for microwave reactions.
Ordering information: Supplied with 20 mm aluminum seals with installed blue PTFE-faced silicone septa. The septa are 0.125" thick (0.005" PTFE; 0.120" silicone).
Customers Who Bought This Also Bought