Nalgene® Sample Vials, Low-Density Polyethylene, with Snap Cap, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
With friction fit, snap closures.
Useful for samples, pH cups, weighing bottles, packing, and other general applications. Will not shatter like polystyrene vials.
Customers Who Bought This Also Bought