Shell Vials, Borosilicate Glass, with TITESEAL® Cap, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Straight-sided shell vials are ideal for storing dry products.
With open-bottom plug-style closures. Manufactured from clear N-51A borosilicate glass.
Customers Who Bought This Also Bought