Short Style Shell Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Straight-sided shell vials are fabricated from clear N-51A borosilicate glass.
Ideal storage containers for dry products. Without caps.
Customers Who Bought This Also Bought