Micro Sample Vial, Borosilicate Glass, with Screw Cap, Wheaton, DWK Life Sciences
# de catalogue 66021-908
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Cap included:Yes
- Cap Size:8-425
- Cat. no.:66021-908
- Supplier No.:225081
Spécifications
A propos de cet article
A multi-purpose glass vial manufactured from Wheaton-33® low extractable borosilicate glass.
Economical when used for scintillation vial. Will contain micro samples up to 2 ml. Dimensions (with cap): 12×38 mm. With metal-foil lined white plastic cap.
Customers Who Bought This Also Bought