Sample Vials, Screw Thread, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
General-purpose, clear and amber borosilicate glass vials.
Volumes range from 0.5 to 11 drams.
Certifications: Meets USP Type I and ASTM E438, Type I, Class A and B requirements.
Ordering information: Available with or without screw caps. Caps are packed separately.
Customers Who Bought This Also Bought