KIMAX® Sample Vials, Borosilicate Glass, with PTFE-Lined Screw Cap, Kimble Chase, DWK Life Sciences
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificats
A propos de cet article
With PTFE-lined solid-top screw caps attached to vials.
The 16 ml size complies with EPA 600 Series Methods. Made of KG-33 borosilicate glass.
Customers Who Bought This Also Bought