KIMAX® Sample Vials, Borosilicate Glass, with Rubber-Lined Screw Cap, Kimble Chase, DWK Life Sciences
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificats
A propos de cet article
Sample vials with rubber-lined solid top screw caps unattached made of borsilicate glass.
- Clear screw thread sample vials
- Closed-top black phenolic closures with rubber liners, unattached
- Economical and time-saving in the lab
Certifications: Low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought