Accessories for DUOSEAL® Pumps
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
DUOSEAL® pump oil recommended for DUOSEAL® vacuum pumps
Exhaust filters trap exhaust oil mist and reduce pump noise. Supplied with housing and element. Replacement elements for filters also listed.
Features low vapor pressure, chemical stability, ideal viscosity, and no additives or inhibitors.
Customers Who Bought This Also Bought