Accessories for Direct Drive Pumps
Fournisseur: Welch Vacuum
Certificats
A propos de cet article
Premium and Gold pump oils are the only oils recommended for use with Welch direct-drive vacuum pumps
Premium pump oil is designed to resist breakdown at the higher rpms and operating temperatures of direct-drive vacuum pumps. Gold pump oil is a synthetic oil designed for excellent resistance when pumping corrosive gases or vapors.
Customers Who Bought This Also Bought