KONTES® Claisen Adapters for Distillation, Kimble Chase, DWK Life sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Provide dual entry into a variety of flasks.
- Useful for reflux with addition-type reactions
- Joints are the same size
Allow temperature measurement at either top outer opening when used in conjunction with a thermometer adapter. Useful for reflux with addition-type reactions.
Compatible with 179700 glass BEVEL-SEAL™ inlet adapters or 179800 PTFE BEVEL-SEAL™ inlet adapters for thermometer use at either top opening.
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought