KIMAX® Connecting Bulb, Kjeldahl, Cylindrical Type, Kimble Chase, DWK Life Sciences
# de catalogue 51150-045
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Ø:55 mm
- Length:120 mm
- Cat. no.:51150-045
- Supplier No.:16040-55
Spécifications
A propos de cet article
Inlet and outlet tubes are curved in opposite directions within the bulb
Lower tubes are 11 to 12mm O.D. to fit special Kjeldahl stoppers.
Customers Who Bought This Also Bought