SP Wilmad-LabGlass Dean-Stark Moisture Traps, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Calibrated to contain at 20°C
For determination of water in various materials by distillation with an immiscible solvent. May be used for ASTM Method D95 and other similar ASTM methods. Traps have a full length [ST] inner joint for vapor tube connection to distilling flask and a [ST] outer joint of the same size for connection to condenser. Traps are graduated in 0.1mL divisions from 0–2mL. Above 2mL, they are graduated in 0.2mL divisions.
Customers Who Bought This Also Bought