SP Wilmad-LabGlass Dean-Stark Moisture Traps with Additional Line, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
These distilling receivers are provided with an additional solvent return line which avoids recycling condensate to the flask.
- Borosilicate glass
With full length Standard Taper inner joint for vapor tube connection to distilling flask and a Standard Taper outer joint of the same size for connection to condenser.
Customers Who Bought This Also Bought