Multi-Compartment Glove Dispensers, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Three- and four-compartment glove dispensers are made from clear, clean, 6mm (1/4") thick acrylic or PETG and feature a separate lid for each compartment
Edges are flame-treated for smoothness. Supplied with a heavy-duty wall bracket and support bar for mounting dispenser to a wall.
Customers Who Bought This Also Bought