Sterile Glove Dispensers, Free-Standing, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Designed to conveniently dispense sterile gloves
Dispensers maximize space utilization and feature first in/first out glove usage. Convenient to fill and maintain. Constructed of durable 6mm (1/4") thick acrylic or PETG. Free-standing; wall-mountable dispensers are sold separately.
Dispensers maximize space utilization and feature first in/first out glove usage. Convenient to fill and maintain. Constructed of durable 6mm (1/4") thick acrylic or PETG. Free-standing; wall-mountable dispensers are sold separately. Dimensions: 17.2W x 43.1H x 30.2D cm (7 x 17 x 12").
Protect gloves from ozone degradation
Customers Who Bought This Also Bought