WHEATON® E-Z Crimper™ Hand-Operated Crimpers, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Crimpers feature polished crimping jaws for consistent sealing, a redesigned spring assembly, and a short, cushioned ergonomic handle that helps reduce hand fatigue and adds comfort
Note: Autoclaving may change the appearance or reduce product life.
An adjustable stop allows tightening of the seal while maintaining consistent crimping. The 8, 11, and 13mm models feature tapered jaws for easier vial access in rack. Each crimper size is labeled for quick identification. Autoclavable at 121°C (250°F).
Ordering information: Crimpers work with the Crimpmaster® crimping station. Crimper 80078-992 is supplied with an O-ring to provide the option of crimping the top of a vial to create an extra tight seal. This option is recommended only for standard-opening vials.
Customers Who Bought This Also Bought