A propos de cet article
Synthetic owl pellets provide a safe, realistic, and reusable dissection experience, featuring hand-crafted replicas of skeletons and carefully imitated contents that closely mimic natural owl pellets.
- A clean and safe dissection experience
- Realistic replica of natural owl pellet
- Handmade
- Reusable
Our simulated Owl pellets offer a hands-on approach to learning the intricacies of food webs and ecosystems through safe, realistic, and engaging replicas. Perfect for schools and colleges looking to enrich their biology or ecology curriculum, these simulated pellets provide a unique insight into the food chain and predator-prey relationships without the mess or safety concerns of real owl pellets. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Dimensions: 3×1.5×1.5"
Weight: 0.2 lbs.