Vial Crushers, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Unit breaks and separates liquid from vials and ampules for environmentally responsible waste handling.
- Reduces overall waste disposal costs by reducing the total waste volume
- Accomodates vials from 1 ml to 100 ml
- Can handle approximately 5000×2 ml vials per hour
Safe to use in a fume hood or to connect to a vacuum system to limit exposure to flammable or toxic vapors. Crushed glass is automatically separated from solvent for separate disposal.
Customers Who Bought This Also Bought