Cleanroom Dispenser with One Compartment, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Designed to protect gloves, bouffants, shoe covers, etc
Easy access and assures first in/first out usage.
Easy access and assures first in/first out usage.
Wall mounts or free-standing/wall mount bracket on back.
Flame polished edges for smoothness.
Unique dispensing port provides.
Spécifications
- Catalog No:
- 89108-030
- 98106-902
- PPE type:
- Multi-use
- Multi-use
- Material:
- Acrylic
- Acrylic
- No. of compartments:
- 1
- 1
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable