Dispenser Storage Cabinet, Bandy
# de catalogue 98000-554
Fournisseur: Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Dispenser type:Floor-standing
- No. of compartments:8
- PPE type:Multi-use
- Description:Storage dispenser, stainless steel
- Material:Stainless steel
- W×D×H:127×61×177.8 cm (50×24×70")
- Supplier No.:CBN02011
Spécifications
A propos de cet article
Cabinet features eight see-through compartments with Plexiglas® doors
Constructed of Type 304 stainless steel with a No. 4 finish.
Customers Who Bought This Also Bought