Bouffant Cap Dispenser, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
Bouffant cap dispensers are designed to provide safe and convenient storage and access to large volumes of caps, beard covers, or similarly packaged garments.
- Dispensers can be wall-mounted or used free-standing
- Edges are flame-treated for smoothness
All dispensers are equipped with a full-length hinged lid for quick and easy interior access. Available in either acrylic or PETG construction, all units have a thickness of 3 mm (1/8").
Spécifications
- Catalog No:
- 10693-140
- 89208-666
- 98106-944
- 10693-138
- PPE type:
- Bouffant caps
- Bouffant caps
- Bouffant caps
- Bouffant caps
- Material:
- PETG
- PETG
- PETG
- PETG
- No. of compartments:
- 1
- 1
- 1
- 1
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Thickness:
- 3 mm
- 3 mm
- 3 mm
- 3 mm