Apparel Dispensers, S-Curve Technologies
Fournisseur: S-Curve Technologies
Certificats
A propos de cet article
One-compartment apparel dispensers are designed to dispense aprons, coveralls, frocks, and smocks
They feature an open front for easy access and are made of durable clear acrylic or PETG with a thickness of 6 mm (¹/₄"). Dispensers can be mounted to the wall or used free-standing. Edges are flame-treated for smoothness. Available with or without a lid.
Ordering information: Supplied with heavy-duty mounting bracket and support bar. Dimensions: 61 x 38.5 x 26 cm (24 x 15.5 x 10.5")
Spécifications
- Catalog No:
- 14007-126
- 89208-660
- PPE type:
- Multi-use
- Multi-use
- Material:
- Acrylic
- Acrylic
- No. of compartments:
- 1
- 1
- Dispenser type:
- Floor-standing/wall-mountable
- Floor-standing/wall-mountable
- Thickness:
- 6 mm (¹/₄")
- 6 mm (¹/₄")