VWR® Critical Print™ Cleanroom Spiral Notebooks
Supplier: VWR International
Certificates
About this item
Notebooks feature semi-transparent, high-density polyethylene covers that withstand harsh chemical vapors.
- Low particulate level
- Semi-transparent polyethylene cover
Low-particulate, 104g/m2 (28#) bond paper. Each notebook contains 50 sheets.
Customers Who Bought This Also Bought