WHEATON® Polypropylene Crimp-Top/Snap-Cap Specialty Vial, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
This lightweight, economical vial can be used in most standard gas and liquid chromatography autosamplers.
Conical interior design ensures maximum retrieval of contents and no time-consuming assembly of vials or limited-volume inserts. Constructed of polypropylene for reliable chemical resistance.
Customers Who Bought This Also Bought