About this item
Economical wiping seal dispenser that can handle many of the fluids used in biological labs
- Economy dispenser for bases, buffers and other liquids used in biology labs
- Replaceable dispensing cartridge for long service life
- Not for use with scintillation fluid
Caution: Dispensers are not for use with scintillation fluid, hydrofluoric acid, concentrated or oxidizing acids, or nonpolar solvents.
Buffers, bases, culture media, and even sodium azide solutions can all be dispensed easily. These dispensers precisely dispense measured volumes of weak acids, alkaline solutions, polar solvents, isotonic solutions, methanol, and acetylacetone. The notched volume selector enables quick and exact volume adjustment. The spring-driven refill function provides semi-automatic operation. Dispensers can be easily disassembled to replace parts and extend instrument life. Sterile dispensing can be performed with additional accessories.