VWR® Talon® Aluminum Support Plate
# de catalogue 60079-000
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Ø:152 mm (6")
- Cat. no.:60079-000
Spécifications
A propos de cet article
Talon® Aluminum Support Plate fits standard lattice lab frames using VWR® Talon® connectors 21572-501, 21572-556, or 60097-055.
- Aluminum construction
- Compatible with standard lattice lab frames
- Hold a variety of media vessels
Supports beakers, mantles, and Petri dishes.
Customers Who Bought This Also Bought